Learn More
Invitrogen™ RASSF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592352
Description
Immunogen sequence: MTMMAHQYPS WIFINEKTFI TREQLNSLLK TYNIFYENQK NLHILYGETE DGKLIVEGML DIFWGVKRPI QLKIQDEKPF SSFTSMKSSD VFSSKGMTRW GEFDDLYRIS ELDRTQIPMS EKRNSQEDYL SYHSNTLKPH AKDEPDSPVL YRTMSEAALV RKRMKPLMMD RKERQKNRAS INGHFYNHEI ENSPQDFALH IIFATGEQRR LKKTDIPLLQ RLLQGPSEKN ARIFLMDKDA EEISSDVAQY INFHFSLLES ILQRLNEEEK REIQRIVTKF NKEKAIILKC LQNKLVIKTE TTV; Positive Samples: Mouse intestine, Rat kidney.
RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.
Specifications
| RASSF6 | |
| Polyclonal | |
| Unconjugated | |
| Rassf6 | |
| 1600016B17Rik; DKFZp686K23225; putative RAS binding protein; Ras association (RalGDS/AF-6) domain family 6; Ras association (RalGDS/AF-6) domain family member 6; Ras association domain family member 6; ras association domain-containing protein 6; RASSF6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 305251, 73246 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4QR82, Q80UQ2 | |
| Rassf6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-303 of human RASSF6 (NP_0012573201). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.