missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF6 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81798UV
This item is not returnable.
View return policy
Description
RASSF6 Polyclonal antibody specifically detects RASSF6 in Human samples. It is validated for Western Blot, ELISA
Specifications
| RASSF6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 350 | |
| DKFZp686K23225, putative RAS binding protein, Ras association (RalGDS/AF-6) domain family 6, Ras association (RalGDS/AF-6) domain family member 6, ras association domain-containing protein 6 | |
| RASSF6 antibody was raised against a 14 amino acid synthetic peptide near the center of human RASSF6. The immunogen is located within amino acids 230 - 280 of RASSF6. Amino Acid Squence: LQKFKIENSPQDFA | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, GPCR, Signal Transduction | |
| 166824 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction