missing translation for 'onlineSavingsMsg'
Learn More

RASSF6 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18417485 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417485 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417485 Supplier Novus Biologicals Supplier No. NBP281798AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RASSF6 Polyclonal antibody specifically detects RASSF6 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF6
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias DKFZp686K23225, putative RAS binding protein, Ras association (RalGDS/AF-6) domain family 6, Ras association (RalGDS/AF-6) domain family member 6, ras association domain-containing protein 6
Host Species Rabbit
Immunogen RASSF6 antibody was raised against a 14 amino acid synthetic peptide near the center of human RASSF6. The immunogen is located within amino acids 230 - 280 of RASSF6. Amino Acid Squence: LQKFKIENSPQDFA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 166824
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.