missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35486-100ul
This item is not returnable.
View return policy
Description
RASSF4 Polyclonal antibody specifically detects RASSF4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RASSF4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| AD037, MGC44914, Ras association (RalGDS/AF-6) domain family 4, Ras association (RalGDS/AF-6) domain family member 4, Ras association domain family 4, ras association domain-containing protein 4, tumor suppressor RASSF4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human RASSF4 (NP_114412.2).,, Sequence:, MKEDCLPSSHVPISDSKSIQKSELLGLLKTYNCYHEGKSFQLRHREEEGTLIIEGLLNIAWGLRRPIRLQMQDDREQVHLPSTSWMPRRPSCPLKEPSPQNGNITAQGPSIQPVHKAESSTDSSGPLEEAEEAPQLMRTKSDASCMSQRRPKCRAPGEAQ | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 83937 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction