missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-20967
This item is not returnable.
View return policy
Description
RASSF3 Polyclonal specifically detects RASSF3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| RASSF3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| MGC119194, MGC119195, MGC119197, Ras association (RalGDS/AF-6) domain family 3, Ras association (RalGDS/AF-6) domain family member 3, ras association domain-containing protein 3, RASSF5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:2000 | |
| NP_835463.1 | |
| RASSF3 | |
| Peptide with sequence CKPPQTSPNSGK corresponding to C-Terminus according to NP_835463.1. | |
| 0.1 mg | |
| Cancer | |
| 283349 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction