missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASSF2 Recombinant Rabbit Monoclonal Antibody (23GB4605), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | RASSF2 |
|---|---|
| Clone | 23GB4605 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089141
![]() |
Invitrogen™
MA552712 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088335
|
Invitrogen™
MA552713 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RASSF2 Recombinant Monoclonal antibody specifically detects RASSF2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RASSF2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P50749 | |
| 9770 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4605 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1 | |
| A synthesized peptide derived from human RASSF2 (301-326AA). | |
| Primary | |
| -20°C | |
| RASSF2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title