missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASSF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103099
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RASSF2.
Potential tumor suppressor. Acts as a KRAS-specific effector protein. May promote apoptosis and cell cycle arrest. Stabilizes STK3/MST2 by protecting it from proteasomal degradation.
Specifications
| RASSF2 | |
| Polyclonal | |
| Unconjugated | |
| RASSF2 | |
| 3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 215653, 311437, 9770 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P50749, Q3B7D5, Q8BMS9 | |
| RASSF2 | |
| A synthesized peptide derived from human RASSF2(Accession P50749), corresponding to amino acid residues V112-R162. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction