missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASSF2 Monoclonal Antibody (1509CT269.10.9.29)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524746
This item is not returnable.
View return policy
Description
RASSF2 Monoclonal Antibody for Western Blot, Flow
Potential tumor suppressor. Acts as a KRAS-specific effector protein. May promote apoptosis and cell cycle arrest. Stabilizes STK3/MST2 by protecting it from proteasomal degradation.
Specifications
| RASSF2 | |
| Monoclonal | |
| 1.46 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P50749 | |
| RASSF2 | |
| Recombinant protein between 1-326 amino acids from human RASSF2. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Western Blot | |
| 1509CT269.10.9.29 | |
| Unconjugated | |
| RASSF2 | |
| 3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 9770 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction