missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41130
This item is not returnable.
View return policy
Description
RASSF10 Polyclonal specifically detects RASSF10 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RASSF10 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| RASSF10 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human RASSF10. The immunogen is located within amino acids 50 - 100 of RASSF10. Amino Acid Squence: HGPGELPEPPNEDDEDD | |
| 0.1 mg | |
| Primary | |
| RASSF10 antibody will not cross-react with other RAS association domain-containing family members. | |
| Store at 4°C. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 ug/mL ELISA, Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL | |
| Peptidylglycine Alpha-Amidating Monooxygenase COOH-Terminal Interactor-Like, Ras Association (RalGDS/AF-6) Domain Family (N-Terminal) Member 10, Ras Association Domain-Containing Protein 10 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 644943 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction