missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASSF1 Antibody (3F3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04342AF594
This item is not returnable.
View return policy
Description
RASSF1 Monoclonal specifically detects RASSF1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| RASSF1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RASSF1 | |
| Recombinant human RASSF1A (1-340 aa), purified from E. coli (NP_009113). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3F3 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5 | |
| Mouse | |
| Protein G purified | |
| Breast Cancer, Cell Cycle and Replication, Mitotic Regulators | |
| 11186.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction