missing translation for 'onlineSavingsMsg'
Learn More

Raspberry ketone, MedChemExpress

Product Code. 16479156
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479156 Supplier Medchem Express Supplier No. HYN1426

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

MedChemExpress Raspberry ketone is a major aromatic compound of red raspberry, widely used as a fragrance in cosmetics and as a flavoring agent in foodstuff; also shows PPAR-α agonistic activity.

Raspberry ketone is a major aromatic compound of red raspberry, widely used as a fragrance in cosmetics and as a flavoring agent in foodstuff

Specifications

Chemical Name or Material Raspberry ketone
CAS 5471-51-2
Color White
Purity Grade Notes Research
Molecular Formula C10H12O2
Quantity 100 mg
Synonym Frambione 4-(4-Hydroxyphenyl)-2-butanone
Solubility Information DMSO : 100 mg/mL (609.01 mM; Need ultrasonic)
SMILES CC(CCC1=CC=C(O)C=C1)=O
Molecular Weight (g/mol) 164.2
Formula Weight 164.2
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.