missing translation for 'onlineSavingsMsg'
Learn More

Raspberry Ketone Glucoside, TRC

Product Code. 30428039
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428039 500 mg 500mg
30423054 250 mg 250mg
30421420 2.5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428039 Supplier Toronto Research Chemicals Supplier No. TRCR126043500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38963-94-9
Molecular Formula C16H22O7
Molecular Weight (g/mol) 326.34
IUPAC Name 4-[4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyphenyl]butan-2-one
SMILES CC(=O)CCc1ccc(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H22O7/c1-9(18)2-3-10-4-6-11(7-5-10)22-16-15(21)14(20)13(19)12(8-17)23-16/h4-7,12-17,19-21H,2-3,8H2,1H3/t12-,13-,14+,15-,16-/m1/s1
Formula Weight 326.1366
Chemical Name or Material Raspberry Ketone Glucoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.