missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASGRP3 Monoclonal Antibody (C.491.8)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514900
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation.
Specifications
| RASGRP3 | |
| Monoclonal | |
| 2 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q8IV61 | |
| Rasgrp3 | |
| Synthetic peptide corresponding to residues near the carboxy teminus of human RasGRP3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| C.491.8 | |
| Unconjugated | |
| Rasgrp3 | |
| BC066069; calcium and DAG-regulated guanine nucleotide exchange factor III; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor III; CalDAG-GEFIII; Gm327; GRP3; Guanine nucleotide exchange factor for Rap1; KIAA0846; OTTHUMP00000201245; RAS guanyl releasing protein 3; RAS guanyl releasing protein 3 (calcium and DAG-regulated); ras guanyl-releasing protein 3; RAS, guanyl releasing protein 3; Rasgrp3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 240168, 25780 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction