missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38466-20ul
This item is not returnable.
View return policy
Description
RASGRP3 Polyclonal antibody specifically detects RASGRP3 in Human,Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| RASGRP3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Calcium and DAG-regulated guanine nucleotide exchange factor III, GRP3KIAA0846ras guanyl-releasing protein 3, Guanine nucleotide exchange factor for Rap1, RAS guanyl releasing protein 3 (calcium and DAG-regulated) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 540-689 of human RASGRP3 (NP_056191.1).,, Sequence:, VLACRRFARAPSLSSGHGSLPGSPSLPPAQDEVFEFPGVTAGHRDLDSRAITLVTGSSRKISVRLQRATTSQATQTEPVWSEAGWGDSGSHTFPKMKSKFHDKAAKDKGFAKWENEKPRVHAGVDVVDRGTEFELDQDEGEETRQDGEDG | |
| 20 μL | |
| Phospho Specific | |
| 25780 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction