missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20089
This item is not returnable.
View return policy
Description
RASGRP2 Polyclonal specifically detects RASGRP2 in Human samples. It is validated for Western Blot.
Specifications
| RASGRP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| Calcium and DAG-regulated guanine nucleotide exchange factor I, CALDAG-GEFI, CDC25Lcalcium and diacylglycerol-regulated guanine nucleotide exchange factor I, Cdc25-like protein, F25B3.3 kinase-like protein, guanine exchange factor MCG7, hCDC25L, MCG7, RAS guanyl nucleotide-releasing protein 2, RAS guanyl releasing protein 2 (calcium and DAG-regulated), RAS guanyl-releasing protein 2 | |
| Rabbit | |
| 69 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.63mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q7LDG7 | |
| RASGRP2 | |
| Recombinant protein encompassing a sequence within the center region of human RASGRP2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10235 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction