missing translation for 'onlineSavingsMsg'
Learn More

RASGRP2 Antibody (3D8), Novus Biologicals™

Product Code. 18334599 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334599 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334599 Supplier Novus Biologicals Supplier No. H00010235M09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RASGRP2 Monoclonal antibody specifically detects RASGRP2 in Human, Canine samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3D8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Alias Calcium and DAG-regulated guanine nucleotide exchange factor I, CALDAG-GEFI, CDC25Lcalcium and diacylglycerol-regulated guanine nucleotide exchange factor I, Cdc25-like protein, F25B3.3 kinase-like protein, guanine exchange factor MCG7, hCDC25L, MCG7, RAS guanyl nucleotide-releasing protein 2, RAS guanyl releasing protein 2 (calcium and DAG-regulated), RAS guanyl-releasing protein 2
Host Species Mouse
Immunogen RASGRP2 (NP_005816, 65 a.a. ~ 164 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GTLDLDKGCTVEELLRGCIEAFDDSGKVRDPQLVRMFLMMHPWYIPSSQLAAKLLHIYQQSRKDNSNSLQVKTCHLVRYWISAFPAEFDLNPELAEQIKE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10235
Target Species Human, Canine
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.