missing translation for 'onlineSavingsMsg'
Learn More

Rasgrp1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £470.00

Specifications

Antigen Rasgrp1
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18697448
View Documents
Novus Biologicals
NBP2-68636-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18661468
View Documents
Novus Biologicals
NBP2-68636
100 μg
£470.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rasgrp1 Polyclonal antibody specifically detects Rasgrp1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Rasgrp1
Immunofluorescence
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.2) and 40% Glycerol
10125
IgG
Protein A purified
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Polyclonal
Purified
RUO
Human
RAS guanyl-releasing protein 1, CALDAG-GEFI, CALDAG-GEFII, hRasGRP1, RAS guanyl releasing protein 1 (calcium and DAG-regulated), RASGRP
This antibody was developed against a recombinant protein corresponding to amino acids: RKTAQDTLYVLPSPTSPCPSPVLVRKRAFVKWENKDSLIKSKEELRHLRLPTYQELEQEINTLKADNDALKIQLKYAQKKIESLQLEKSNHVLAQMEQGD
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.