missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rasgrp1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94157-0.1ml
This item is not returnable.
View return policy
Description
Rasgrp1 Polyclonal antibody specifically detects Rasgrp1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Rasgrp1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| RAS guanyl-releasing protein 1, CALDAG-GEFI, CALDAG-GEFII, hRasGRP1, RAS guanyl releasing protein 1 (calcium and DAG-regulated), RASGRP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 598-797 of human RASGRP1 (NP_005730.2). PVAPTENNTSVGPVSNLCSLGAKDLLHAPEEGPFTFPNGEAVEHGEESKDRTIMLMGVSSQKISLRLKRAVAHKATQTESQPWIGSEGPSGPFVLSSPRKTAQDTLYVLPSPTSPCPSPVLVRKRAFVKWENKDSLIKSKEELRHLRLPTYQELEQEINTLKADNDALKIQLKYAQKKIESLQLEKSNHVLAQMEQGDCS | |
| 0.1 mL | |
| Signal Transduction | |
| 10125 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction