missing translation for 'onlineSavingsMsg'
Learn More

RASGRP 4 Antibody (1F3), Novus Biologicals™

Product Code. 18411099 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411099 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411099 Supplier Novus Biologicals Supplier No. H00115727M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RASGRP 4 Monoclonal antibody specifically detects RASGRP 4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRP 4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 1F3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Alias guanyl nucleotide releasing protein 4, RAS guanyl releasing protein 4, RAS guanyl-releasing protein 4
Host Species Mouse
Immunogen RASGRP4 (NP_733748, 574 a.a. ∽ 673 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CGLCCHKHCRDQVKVECKKRPGAKGDAGPPGAPVPSTPAPHASCGSEENHSYTLSLEPETGCQLRHAWTQTESPHPSWETDTVPCPVMDPPSTASSKLDS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 115727
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.