missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRF1 Polyclonal Antibody

RASGRF1 Polyclonal Antibody
Brand: Invitrogen PA5102974
This item is not returnable.
View return policy
Description
RASGRF1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| RASGRF1 | |
| Polyclonal | |
| Unconjugated | |
| RASGRF1 | |
| AI844718; ASPP1; Cdc25; CDC25L; CDC25Mm; Gnrp; GRF1; GRF55; Grfbeta; guanine nucleotide exchange factor; guanine nucleotide-releasing factor 1; guanine nucleotide-releasing factor, 55 kD; guanine nucleotide-releasing protein; H-GRF55; KIAA0771; P140 Ras-GRF; p190; P190-A; p190RhoGEF; PP13187; PPP1R13B; Ras guanine release factor beta; Ras protein specific guanine nucleotide releasing factor 1; RAS protein-specific guanine nucleotide-releasing factor 1; Rasgrf1; ras-GRF1; Ras-specific guanine nucleotide-releasing factor 1; Ras-specific guanine nucleotide-releasing factor, CDC25 homolog; ras-specific nucleotide exchange factor CDC25 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5923 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13972 | |
| RASGRF1 | |
| A synthesized peptide derived from human RASGRF1(Accession Q13972), corresponding to amino acid residues Q2-F52. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction