missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRF1 Antibody (S06-5H5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19949-100ul
This item is not returnable.
View return policy
Description
RASGRF1 Monoclonal antibody specifically detects RASGRF1 in Human, Rat samples. It is validated for Western Blot
Specifications
| RASGRF1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| S06-5H5 | |
| Unconjugated | |
| CDC25guanine nucleotide exchange factor, CDC25L, GNRPguanine nucleotide-releasing factor 1, GRF1, GRF55, guanine nucleotide-releasing factor, 55 kD, Guanine nucleotide-releasing protein, H-GRF55, PP13187, Ras protein-specific guanine nucleotide-releasing factor 1, ras-GRF1, ras-specific guanine nucleotide-releasing factor 1, Ras-specific guanine nucleotide-releasing factor, CDC25 homolog, Ras-specific nucleotide exchange factor CDC25 | |
| A synthetic peptide of human RASGRF1 (Uniprot # Q13972) | |
| 100 μg | |
| Signal Transduction | |
| 5923 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction