missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASGRF1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95233-0.02ml
This item is not returnable.
View return policy
Description
RASGRF1 Polyclonal antibody specifically detects RASGRF1 in Mouse samples. It is validated for Western Blot
Specifications
| RASGRF1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| CDC25guanine nucleotide exchange factor, CDC25L, GNRPguanine nucleotide-releasing factor 1, GRF1, GRF55, guanine nucleotide-releasing factor, 55 kD, Guanine nucleotide-releasing protein, H-GRF55, PP13187, Ras protein-specific guanine nucleotide-releasing factor 1, ras-GRF1, ras-specific guanine nucleotide-releasing factor 1, Ras-specific guanine nucleotide-releasing factor, CDC25 homolog, Ras-specific nucleotide exchange factor CDC25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human RASGRF1 (NP_001139120.1). MQKGIRLNDGHVASLGLLARKDGTRKGYLSKRSSDNTKWQTKWFALLQNLLFYFESDSSSRPSGLYLLEGCVCDRAPSPKPALSAKEPLEKQHYFTVNFSHENQKALELRTEDAKDCDEWVAAIAHASYRTLATEHEALMQKYLHLLQIVETEKTVAKQLRQQIEDGEIEIERLKAEITSLLKDNERIQS | |
| 0.02 mL | |
| Signal Transduction | |
| 5923 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction