missing translation for 'onlineSavingsMsg'
Learn More

RASGRF1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18364673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364673 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364673 Supplier Novus Biologicals Supplier No. NBP306655AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RASGRF1 Polyclonal antibody specifically detects RASGRF1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRF1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CDC25guanine nucleotide exchange factor, CDC25L, GNRPguanine nucleotide-releasing factor 1, GRF1, GRF55, guanine nucleotide-releasing factor, 55 kD, Guanine nucleotide-releasing protein, H-GRF55, PP13187, Ras protein-specific guanine nucleotide-releasing factor 1, ras-GRF1, ras-specific guanine nucleotide-releasing factor 1, Ras-specific guanine nucleotide-releasing factor, CDC25 homolog, Ras-specific nucleotide exchange factor CDC25
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RASGRF1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5923
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.