missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASGEF1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560758
This item is not returnable.
View return policy
Description
Immunogen sequence: RDERMMRNLK DLAHRIASGE ETYRKNVQQM MQCLIRKLAA LSQYEEVLAK ISSTSTDRLT VLKTKPQSIQ RDIITVC Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.
Guanine nucleotide exchange factor (GEF) with specificity for RAP2A, it doesn't seems to activate other Ras family proteins (in vitro).
Specifications
| RASGEF1B | |
| Polyclonal | |
| Unconjugated | |
| Rasgef1b | |
| 4732452O09Rik; GPI gamma-4; Gpig4; GPI-gamma 4; GPIgamma4; RasGEF domain family member 1B; RasGEF domain family, member 1B; ras-GEF domain-containing family member 1B; RASGEF1B; RGD1562220 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 153020 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q0VAM2 | |
| Rasgef1b | |
| Recombinant protein corresponding to Human RASGEF1B. Recombinant protein control fragment (Product #RP-96660). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction