missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASGEF1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531598
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (91%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Guanine nucleotide exchange factor (GEF) with specificity for RAP2A, KRAS, HRAS, and NRAS (in vitro). Plays a role in cell migration.
Specifications
| RASGEF1A | |
| Polyclonal | |
| Unconjugated | |
| RASGEF1A | |
| 6330404M18Rik; AI835194; CG4853; CG4853 gene product; RasGEF domain family member 1A; RasGEF domain family, member 1A; ras-GEF domain-containing family member 1A; Rasgef1a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221002 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8N9B8 | |
| RASGEF1A | |
| Recombinant protein encompassing a sequence within the center region of human RASGEF1A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction