Learn More
RASEF Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28162-1-AP-150UL
 View more versions of this product
Product Code. 16876025
  • £349.00 / 150µL
Description
Binds predomitly GDP, and also GTP.
Specifications
Specifications
RASEF
Polyclonal
Unconjugated
RASEF
FLJ31614, RAB45, Ras related protein Rab 45, RASEF
Rabbit
Antigen Affinity Chromatography
RUO
158158, 298138
-20°C
Liquid
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IZ41
RASEF
RASEF Fusion Protein Ag28049
150 μL
Primary
Human, Rat