missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RASEF Polyclonal specifically detects RASEF in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | RASEF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | FLJ31614, RAB45, RAB45, member RAS oncogene family, RAS and EF hand domain containing, RAS and EF-hand domain containing, ras and EF-hand domain-containing protein, Ras-related protein Rab-45 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human RASEF (NP_689786). Peptide sequence AQDKAAMQLSELEEEMDQRIQAAEHKTRKDEKRKAEEALSDLRRQYETEV |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?