missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASEF Antibody (JE65-33), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32886
This item is not returnable.
View return policy
Description
RASEF Monoclonal antibody specifically detects RASEF in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| RASEF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 | |
| FLJ31614, RAB45, RAB45, member RAS oncogene family, RAS and EF hand domain containing, RAS and EF-hand domain containing, ras and EF-hand domain-containing protein, Ras-related protein Rab-45 | |
| Synthetic peptide within human RASEF aa 81-130/740. (Uniprot: Q8IZ41) | |
| 100 μL | |
| Signal Transduction | |
| 158158 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE65-33 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction