missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASEF Antibody (2D5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00158158-M01
This item is not returnable.
View return policy
Description
RASEF Monoclonal antibody specifically detects RASEF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| RASEF | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| FLJ31614, RAB45, RAB45, member RAS oncogene family, RAS and EF hand domain containing, RAS and EF-hand domain containing, ras and EF-hand domain-containing protein, Ras-related protein Rab-45 | |
| RASEF (AAH23566, 631 a.a. ∽ 740 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NIREWVDMIEDAAHETVPIMLVGNKADIRDTAATEGQKCVPGHFGEKLAMTYGALFCETSAKDGSNIVEAVLHLAREVKKRTDKDDSRSITNLTGTNSKKSPQMKNCCNG | |
| 0.1 mg | |
| Signal Transduction | |
| 158158 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| 2D5 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| AAH23566 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction