missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASD2 Monoclonal Antibody (OTI1G1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525305
This item is not returnable.
View return policy
Description
RASD2 Monoclonal Antibody for Flow, Western Blot
Rhes, also known as tumor endothelial marker 2 (TEM2), is a small GTP-binding protein that is predominantly expressed in the striatal region of the brain. This protein belongs to the RASD subfamily of Ras-related GTP-binding protein superfamily and is thought to play a role in the normal development and function of the brain as mice lacking this gene showed increased anxiety levels and motor coordination deficits. Rhes was identified as TEM2 through analysis of genes whose expression was upregulated in tumor endothelium. Tumor endothelial markers are significantly up-regulated during angiogenesis and neoangiogenesis that are crucial for the growth of solid tumors. TEMs localized on the cell surface and conserved across species are of particular interest for future development of anti-angiogenic therapies.
Specifications
| RASD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96D21 | |
| RASD2 | |
| Full length human recombinant protein of RASD2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot | |
| OTI1G1 | |
| Unconjugated | |
| RASD2 | |
| 4930526B11Rik; AU045414; GTP-binding protein Rhes; MGC:4834; Ras homolog enriched in striatum; RASD family member 2; RASD family, member 2; Rasd2; Rhes; SE6C; TEM2; TEM-2; Tumor endothelial marker 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23551, 481284 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction