missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RASD2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77313
This item is not returnable.
View return policy
Description
RASD2 Polyclonal antibody specifically detects RASD2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RASD2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| GTP-binding protein Rhes, MGC:4834, RASD family, member 2, Rhes, TEM2Ras homolog enriched in striatum, Tumor endothelial marker 2 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of the human TEM2. The immunogen is located within amino acids 20 - 70 of TEM2. Amino Acid Squence: FLNGRFEDQYTPTIED | |
| 0.1 mg | |
| Signal Transduction | |
| 23551 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/ Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Q96D21 | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction