missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561617
This item is not returnable.
View return policy
Description
Immunogen sequence: TKENVDVPLV ICGNKGDRDF YREVDQREIE QLVGDDPQRC AYFE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Small GTPase. Negatively regulates the transcription regulation activity of the APBB1/FE65-APP complex via its interaction with APBB1/FE65.
Specifications
| RASD1 | |
| Polyclonal | |
| Unconjugated | |
| RASD1 | |
| Activator of G-protein signaling 1; AGS1; Dexamethasone-induced Ras-related protein 1; DEXRAS1; DEXRAS1 (Dexras1); MGC:26290; ras related dexamethasone induced 1; RAS, dexamethasone-induced 1; Rasd1; ras-related protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51655 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y272 | |
| RASD1 | |
| Recombinant protein corresponding to Human RASD1. Recombinant protein control fragment (Product #RP-100918). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction