missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RASAL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554211
This item is not returnable.
View return policy
Description
Immunogen sequence: LSVLHSLLWE VVSQLDKATV AKLGPLPRVL ADITKSLTNP TPIQQQLRRF TEHNSSPNVS GSLSSGLQKI FEDPT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.
This gene encodes a protein that contains the GAP-related domain (GRD), a characteristic domain of GTPase-activating proteins (GAPs). GAPs function as activators of Ras superfamily of small GTPases. The protein encoded by this gene is able to complement the defective RasGAP function in a yeast system. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Specifications
| RASAL2 | |
| Polyclonal | |
| Unconjugated | |
| RASAL2 | |
| 6530401P13; A330066M24Rik; NGAP; Ras GTPase-activating protein nGAP; Ras protein activator like 1; RAS protein activator like 2; RAS protein activator-like 2; RASAL2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 226525, 9462 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UJF2 | |
| RASAL2 | |
| Recombinant protein corresponding to Human RASAL2. Recombinant protein control fragment (Product #RP-92397). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction