missing translation for 'onlineSavingsMsg'
Learn More

RASAL2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18419063 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419063 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419063 Supplier Novus Biologicals Supplier No. NBP119150AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RASAL2 Polyclonal antibody specifically detects RASAL2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASAL2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias MGC129919, NGAP, ras GTPase-activating protein nGAP, Ras protein activator like 1, RAS protein activator like 2, RAS protein activator-like 2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 888 and 938 of human RAS protein activator like 2 using the numbering given in entry NP_004832.1 (GeneID 9462).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9462
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.