missing translation for 'onlineSavingsMsg'
Learn More

Rasagiline Mesylate, TRC

Product Code. 30009500
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009500 250 mg 250mg
17948684 500 mg 500mg
17929094 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009500 Supplier Toronto Research Chemicals Supplier No. TRCR126000250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 161735-79-1
Molecular Formula C12 H13 N . C H4 O3 S
Molecular Weight (g/mol) 267.34
Synonym (1R)-2,3-Dihydro-N-2-propyn-1-yl-1H-inden-1-amine Methanesulfonate (1:1), 1H-Inden-1-amine, 2,3-dihydro-N-2-propynyl-, (1R)-, methanesulfonate (9CI), 1H-Inden-1-amine, 2,3-dihydro-N-2-propynyl-, (R)-, methanesulfonate, Rasagiline mesylate, Rasagiline methanesulfonate, TV 1012, TVP 1012
IUPAC Name methanesulfonic acid;(1R)-N-prop-2-ynyl-2,3-dihydro-1H-inden-1-amine
SMILES CS(=O)(=O)O.C#CCN[C@@H]1CCc2ccccc12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H13N.CH4O3S/c1-2-9-13-12-8-7-10-5-3-4-6-11(10)12;1-5(2,3)4/h1,3-6,12-13H,7-9H2;1H3,(H,2,3,4)/t12-;/m1./s1
Formula Weight 267.0929
Percent Purity >95%
Chemical Name or Material Rasagiline Mesylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.