missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Rasagiline mesylate

Product Code. 18005802
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18005802 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18005802 Supplier Tocris Bioscience™ Supplier No. 4308/50
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective, irreversible MAO-B inhibitor

Selective irreversible MAO-B inhibitor. Displays antiParkinsonian activity; exhibits neuroprotective and antiapoptotic activity against several neurotoxins in cell culture.
TRUSTED_SUSTAINABILITY

Specifications

CAS 161735-79-1
Quantity 50 mg
Molecular Formula C13H17NO3S
Formula Weight 267.34
Purity >99%
Molecular Weight (g/mol) 267.34
Chemical Name or Material Rasagiline mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.