missing translation for 'onlineSavingsMsg'
Learn More

Rasagiline mesylate, MedChemExpress

Product Code. 16452247
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452247 50 mg 50mg
16436506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452247 Supplier Medchem Express Supplier No. HY14605
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rasagiline (R-AGN1135) mesylate is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

Rasagiline (R-AGN1135) mesylate is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

Specifications

Chemical Name or Material Rasagiline mesylate
CAS 161735-79-1
Color White
Purity Grade Notes Research
Molecular Formula C13H17NO3S
Quantity 50 mg
Synonym (R)-AGN1135 mesylateTVP1012 mesylate
Solubility Information H2O : 25 mg/mL (93.51 mM; Need ultrasonic)
SMILES CS(=O)(O)=O.C#CCN[C@H]1C2=CC=CC=C2CC1
Molecular Weight (g/mol) 267.34
Formula Weight 267.34
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.