missing translation for 'onlineSavingsMsg'
Learn More

Rasagiline, MedChemExpress

Product Code. 16462247
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462247 50 mg 50mg
16446506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462247 Supplier Medchem Express Supplier No. HY14605A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rasagiline (R-AGN1135) is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

Rasagiline (R-AGN1135) is a highly potent selective irreversible mitochondrial monoamine oxidase (MAO) inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

Specifications

Chemical Name or Material Rasagiline
CAS 136236-51-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C12H13N
Quantity 50 mg
Synonym (R)-AGN1135 TVP1012
Solubility Information DMSO : 100 mg/mL (583.98 mM; Need ultrasonic)
SMILES C#CCN[C@H]1C2=CC=CC=C2CC1
Molecular Weight (g/mol) 171.24
Formula Weight 171.24
Percent Purity 98.84%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.