missing translation for 'onlineSavingsMsg'
Learn More

RASA3 Antibody (1F11), Novus Biologicals™

Product Code. 18387119 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387119 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387119 Supplier Novus Biologicals Supplier No. H00022821M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RASA3 Monoclonal antibody specifically detects RASA3 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASA3
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1F11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH47242
Gene Alias GAP1(IP4BP), GAP1IP4BP, GAPIII, GTPase activating protein III, inositol 13,4,5-tetrakisphosphate-binding protein, Ins P4-binding protein, Ins(13,4,5)P4-binding protein, MGC46517, MGC47588, ras GTPase-activating protein 3, RAS p21 protein activator 3
Host Species Mouse
Immunogen RASA3 (AAH47242, 725 a.a. ~ 834 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSI
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22821
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.