missing translation for 'onlineSavingsMsg'
Learn More

Ras-GAP Antibody (OTI1H2), DyLight 594, Novus Biologicals™

Product Code. 18407245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407245 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407245 Supplier Novus Biologicals Supplier No. NBP273804DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ras-GAP Monoclonal antibody specifically detects Ras-GAP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ras-GAP
Applications Immunofluorescence
Classification Monoclonal
Clone OTI1H2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CMAVM;CM-AVM;CMAVM1;GAP;p120;p120GAP;p120RASGAP;PKWS;Ras GTPase-activating protein 1;RASA;RASA1;RASGAP
Host Species Mouse
Immunogen Full length recombinant protein of human RAS p21 (NP_002881) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5921
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.