missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ras-GAP Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18406G
This item is not returnable.
View return policy
Description
Ras-GAP Polyclonal antibody specifically detects Ras-GAP in Human, Mouse samples. It is validated for Western Blot
Specifications
| Ras-GAP | |
| Polyclonal | |
| Western Blot | |
| CMAVM;CM-AVM;CMAVM1;GAP;p120;p120GAP;p120RASGAP;PKWS;Ras GTPase-activating protein 1;RASA;RASA1;RASGAP | |
| Recognizes region between residues Between Between 625 and 675.. | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 5921 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido