missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ras-GAP Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18406V
This item is not returnable.
View return policy
Description
Ras-GAP Polyclonal antibody specifically detects Ras-GAP in Human, Mouse samples. It is validated for Western Blot
Specifications
| Ras-GAP | |
| Polyclonal | |
| Western Blot | |
| CMAVM;CM-AVM;CMAVM1;GAP;p120;p120GAP;p120RASGAP;PKWS;Ras GTPase-activating protein 1;RASA;RASA1;RASGAP | |
| Recognizes region between residues Between Between 625 and 675.. | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 5921 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction