Learn More
Invitrogen™ RARS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110272
Description
Immunogen sequence: LNLPPENLIT SISAVPISQK EEVADFQLSV DSLLEKDNDH SRPDIQVQAK RLAEKLRCDT VVSEISTGQR TVNFKINREL LTKTVLQQVI EDGSKYGLKS ELFSGLPQKK IVVEFSSPNV AKKFHVGHLR STIIGNFIAN LKEALGHQVI RINYLGDWGM QFGLLGTGFQ LFGYEEKLQS NPLQHLFEVY VQVNKEAADD KSVAKAAQEF FQRLELGDVQ ALSLWQKFRD LSIEEYIRVY KRLGVYFDEY SGESFYREKS QEVLKLLESK GLLLKTIKGT AVVDLSGNGD PSSICTVMRS DGTSLYATRD LAAAIDRMDK YNFD.
The protein encoded by this gene is an arginyl-tRNA synthetase that is found in the mitochondrial matrix. Defects in this gene are a cause of pontocerebellar hypoplasia type 6 (PCH6).
Specifications
| RARS2 | |
| Polyclonal | |
| Unconjugated | |
| RARS2 | |
| 1500002I10Rik; arginine--tRNA ligase; Arginyl-tRNA synthetase; arginyl-tRNA synthetase 2, mitochondrial; arginyl-tRNA synthetase 2, mitochondrial (putative); ArgRS; DALRD2; dJ382I10.6; PCH6; PRO1992; probable arginine--tRNA ligase, mitochondrial; probable arginyl-tRNA synthetase, mitochondrial; Rars2; RARSL | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109093, 57038 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3U186, Q5T160 | |
| RARS2 | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.