missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARS Mouse anti-Human, Mouse, Rat, Clone: 1C8G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66815-1-IG-20UL
This item is not returnable.
View return policy
Description
The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. ArgRS (Arginyl-tRNA synthetase), also known as RARS or DALRD1, belongs to the class-I aminoacyl-tRNA synthetase family that includes the related proteins, LeuRS, ValRS and IleRS. These proteins are large monomeric proteins and play a major role in catalyzing the aminoacylation of tRNA by their cognate amino acid. ArgRS localizes to the cytoplasm and exists as a monomer but can also associate with other tRNA synthetases and auxiliary proteins to form a multisubunit complex. In the presence of ATP, arginine (Arg) and tRNA, ArgRS joins Arg to tRNA(Arg) at its synthetic active site. Two cytoplasmic forms of ArgRS have been described in mammals, differing by the addition of a 73 amino acid sequence that is required for ArgRS assembly into the multisubunit complex.Specifications
| RARS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P40329, P54136, Q9D0I9 | |
| Rars | |
| RARS Fusion Protein Ag26399 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 1C8G4 | |
| Unconjugated | |
| RARS | |
| Arginine tRNA ligase, arginyl tRNA synthetase, ArgRS, DALRD1, RARS | |
| Mouse | |
| Protein A | |
| RUO | |
| 104458, 287191, 5917 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction