missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RARRES1 Antibody (513509), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB4255JF646
This item is not returnable.
View return policy
Description
RARRES1 Monoclonal specifically detects RARRES1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RARRES1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RARRES1 | |
| E. coli-derived partial recombinant human RARRES1 protein (amino acids 43-294) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| Human | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 513509 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| RAR-responsive protein TIG1, retinoic acid receptor responder (tazarotene induced) 1, retinoic acid receptor responder protein 1, TIG1Tazarotene-induced gene 1 protein | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction