missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RARB Recombinant Rabbit Monoclonal Antibody (K01_1X75)

Product Code. 30830130
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830130 100 μL
30829533 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830130 Supplier Invitrogen™ Supplier No. MA563857

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RARB Recombinant Monoclonal Antibody for Western Blot

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RARB
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1X75
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene RARB
Gene Accession No. P10826
Gene Alias A830025K23; HAP; HBV-activated protein; hepatitis B virus activated protein; MCOPS12; NR1B2; nuclear receptor subfamily 1 group B member 2; RAR beta; Rarb; RAR-b; RAR-beta; RARbeta1; RAR-Beta2; RAR-Beta4; RAR-epsilon; retinoic acid; retinoic acid receptor; retinoic acid receptor beta; retinoic acid receptor, beta; retinoic acid receptor, beta polypeptide; RRB2
Gene Symbols RARB
Host Species Rabbit
Immunogen A synthetic peptide of human Retinoic Acid Receptor beta.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5915
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.