missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAR gamma/NR1B3 Antibody (SR1891), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21974-100ul
This item is not returnable.
View return policy
Description
RAR gamma/NR1B3 Monoclonal antibody specifically detects RAR gamma/NR1B3 in Human, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| RAR gamma/NR1B3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1891 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| NR1B3RARC, Nuclear receptor subfamily 1 group B member 3, RAR-gamma, retinoic acid receptor gamma, retinoic acid receptor, gamma | |
| A synthesized peptide derived from human RAR gamma/NR1B3 (Uniprot #: P13631) | |
| 100 μg | |
| Cancer, Transcription Factors and Regulators | |
| 5916 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction