missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAR gamma-1,2 Recombinant Rabbit Monoclonal Antibody (9K3Y1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555370
This item is not returnable.
View return policy
Description
Immunogen Sequence: MATNKERLFA AGALGPGSGY PGAGFPFAFP GALRGSPPFE MLSPSFRGLG QPDLPKEMAS LSVETQSTSS EEMVPSSPSP PPPPRVYKPC FVCNDKSSGY.
Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predominantly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.
Specifications
| RAR gamma-1,2 | |
| Recombinant Monoclonal | |
| 1.03 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P13631, P18911 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19411, 5916, 685072 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 9K3Y1 | |
| Unconjugated | |
| RARG | |
| RARG | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RARG. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction