missing translation for 'onlineSavingsMsg'
Learn More

RAR alpha/NR1B1 Antibody (1C10), Novus Biologicals™

Product Code. 18392019 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392019 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392019 Supplier Novus Biologicals Supplier No. H00005914M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RAR alpha/NR1B1 Monoclonal antibody specifically detects RAR alpha/NR1B1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAR alpha/NR1B1
Applications Western Blot, ELISA, Proximity Ligation Assay
Classification Monoclonal
Clone 1C10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Proximity Ligation Assay
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000955
Gene Alias alpha polypeptide, RAR-alpha, retinoic acid receptor, alpha
Host Species Mouse
Immunogen RARA (NP_000955, 315 a.a. ~ 424 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QLLPLEMDDAETGLLSAICLICGDRQDLEQPDRVDMLQEPLLEALKVYVRKRRPSRPHMFPKMLMKITDLRSISAKGAERVITLKMEIPGSMPPLIQEMLENSEGLDTLS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5914
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.