missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPSN Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20453-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of a family of proteins that are receptor associated proteins of the synapse. The encoded protein contains a conserved cAMP-dependent protein kinase phosphorylation site, and plays a critical role in clustering and anchoring nicotinic acetylcholine receptors at synaptic sites by linking the receptors to the underlying postsynaptic cytoskeleton, possibly by direct association with actin or spectrin. Mutations in this gene may play a role in postsynaptic congenital myasthenic syndromes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| RAPSN | |
| Polyclonal | |
| Unconjugated | |
| RAPSN | |
| 43 kDa postsynaptic protein, CMS1D, CMS1E, RAPSN, RAPsyn, RING finger protein 205, RNF205 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19400, 5913 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12672, Q13702 | |
| RAPSN | |
| RAPSN Fusion Protein Ag13896 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction